Introductory Protists Slide Set
Introductory Protists Slide Set, 12, Protists
Catalog # 470177-042
Supplier: Avantor
Jump to:
Product Details & Documents
Survey Set Representing the Four Most Studied Protist Phyla
- Identifying features clearly distinguishable
- Rigorous quality control standards
All slides are prepared as wholemounts unless otherwise specified.
Ordering information: Set includes Volvox, Sexual Stage; Protista; Amoeba proteus; Radiolaria Strew; Euglena; Paramecium caudataum; Stentor; Vorticella; Entamoeba coli; Trypanosama lewisi(sm); Balantidium coli,(sm); Plasmodium vivax, Stages (sm).
Specifications
- Type:Protists
- Désignation:Introductory Protists Slide Set
- No. of Slides:12