Introductory Planetarium
Introductory Planetarium
Supplier:   Science First
Introductory Planetarium
Supplier:  Science First
Certificates
Introductory Planetarium
Introductory Planetarium
2 Options Available Below
Product Details & Documents
Introductory Planetarium is a simple model for hands-on demonstrations.
  • Illumination Available
  • Instructions Included
  • Length: 22"
  • Width: 7½"
  • Height: 12"
Operating the hand-driven system of this planetarium causes the Earth to rotate around the Sun. With the available illumination, the model features a lit Sun that focuses a beam of light on Earth as it rotates to help students visualize the process of night and day.
Instructions are included.
Product Family Options
Product Information
  • Désignation
    Width
    Longueur
    Hauteur
    Availability
    Price
  • Introductory Planetarium
    7½"
    22"
    12"
    Log in for availability
    Log in for availability
    Each
    Each - $427.95
    $427.95
    Each
       $427.95
    Supplier Number: 653-3030
  • Introductory Planetarium, With Illumination
    7½"
    22"
    12"
    Log in for availability
    Log in for availability
    Each
    Each - $306.70
    $306.70
    Clearance
    Each
       $306.70
    Supplier Number: 653-3040