Introductory Epithelial Tissues Slide Set
Introductory Epithelial Tissues Slide Set, 5, Animal
Catalog # 470177-706
Supplier: Avantor
Jump to:
Product Details & Documents
Illustrates Classification Types
- Identifying features clearly distinguishable
- Rigorous quality control standards
Ordering information: Set includes Squamous epithelium, frog skin (wm); Simple squamous mesothelium, mesentery (wm); Simple ciliated Columnar epithelium (sect); Stratified squamous epithelium, esophagus (sect); Simple cuboidal epithelium, thyroid (sect).
Spécifications
- Type:Animal
- Désignation:Introductory Epithelial Tissues Slide Set
- No. of Slides:5