Introductory Bacteria Slides
Introductory Bacteria Slides, 10, Monera
Catalog # 470177-040
Supplier: Avantor
Jump to:
Product Details & Documents
Compare and contrast bacterial morphology
- Identifying features clearly distinguishable
- Rigorous quality control standards
Slides are supplied in a durable storage box.
: Set includes Azotobacter chroococcum (sm); Spirillum volutans (sm); Agrobacterium tumefaciens (sm); Bacillus anthracis (sm); Escherichia coli (sm); Mycobacterium tuberculosis (sm); Salmonella enteritidis (sm); Staphylococcus aureus (sm); Treponema sp., (sm); Root nodules (cs).
Spécifications
- Type:Monera
- Désignation:Introductory Bacteria Slides
- No. of Slides:10