ContactUsLinkComponent
Kemtec® Introduction to Toxicology
Catalogue # 470144-334
Fournisseur:  Ward's Science
Kemtec® Introduction to Toxicology
Catalogue # 470144-334
Fournisseur:  Ward's Science
Supplier Number:  10-530
Spécifications
  • Description:
    Introduction to toxicology
  • No. of Lab Groups:
    6
  • Cat. no.:
    470144-334
  • Supplier No.:
    10-530
Spécifications
À propos de cet article
Learn how to screen for drugs.
  • Practice thin layer chromatography
  • Learn about polarity and pH
  • Time required: 50 minute class period
Acquaint your students with the variety of procedures used in testing unknowns against known standards for the presence of narcotics. Techniques employed include TLC, testing polarity/pH of molecules, urinalysis, and more. The activity comes with enough materials for six setups and a teacher and student manual.
Ordering information: Due to the nature of this item, it cannot be shipped by air under any circumstances. Please select ground shipping at time of checkout.
Delivery: See datasheet for contents information.