Introduction to Physical Properties of Proteins Lab Activity
Catalogue # 470008-064
Fournisseur: Kemtec
Spécifications
- Description:Introduction to Physical Properties of Proteins Lab Activity
- Cat. no.:470008-064
- Supplier No.:8-152
Spécifications
À propos de cet article
Provide a Thorough Overview of the Function of Proteins
Students will discover how physical parameters, such as temperature, pH, salt concentration and dielectric constants affect the isolation and purification of proteins. They will conduct experiments in which they will precipitate casein out of milk, separate albumin and gelatin through fractionation, determine the effects of acetone on albumin and gelatin, denature albumin, and precipitate protein with lead acetate. It includes enough materials for eight setups, a teacher’s guide, and student data sheets.