ContactUsLinkComponent
Introduction to Conductivity Experiment Kit (Refill Kit Only)
Catalogue # 470003-766
Fournisseur:  Lab-Aids
undefined
Introduction to Conductivity Experiment Kit (Refill Kit Only)
Catalogue # 470003-766
Fournisseur:  Lab-Aids
Supplier Number:  310
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Spécifications
  • Description:
    Introduction to Conductivity Experiment Kit
  • Cat. no.:
    470003-766
  • Supplier No.:
    310
Spécifications
À propos de cet article
With this kit students study the principles of conductors and insulators; study and develop the principles of electrolytes and non-electrolytes using acids, bases, salts, alcohols, and hydrocarbons; test a variety of common materials for their conductivity characteristics; and learn to differentiate between strong, weak, and non-conductors of electricity.
Ordering information: Three 9 V batteries required for conductivity indicators, not included. Includes 3 LED conductivity indicators; 15 Lab-Aids® Chemplates® with spatula; 3 drop controlled btls deionized water; 2 drop controlled btls each 6 M hydrochloric acid, denatured alcohol, glacial acetic acid; 1 vial each calcium hydroxide, sodium chloride crystals, sucrose crystals; 1 pkg. each copper wire with insulation, metal paper clips, plastic toothpicks, rubber bands, cut string; 30 student guides; 1 instruction manual with MSDS. Note: Only the refill kit is available at this time.