Intercellular Bridges Slide
Intercellular Bridges, Section, Iron Hematoxylin
470182-608
: Avantor
Illustrates how Cells are Connected
- Identifying features clearly distinguishable
- Rigorous quality control standards
Shown within squamous epithelium.
- Désignation:Intercellular Bridges
- Stain:Iron Hematoxylin
- Orientation:Section