Insta-Check pH Papers
Supplier: Micro Essential
Jump to:
À propos de cet article
The Ultimate in Wide Range, General Purpose pH Papers.
- One dip determines any pH from 0 - 13
- Distinct and easily read color separations
Insta-Check 0 - 13 provides 14 brilliant colors on 1 paper giving instant pH checks over the entire range with accurate, reproducible readings; and immediate response. Available in different widths to meet your special requirements and preferences. Paper rolls come with dispenser; refills do not.
3 Options Available Below
Product Details & Documents
The Ultimate in Wide Range, General Purpose pH Papers.
- One dip determines any pH from 0 - 13
- Distinct and easily read color separations
Insta-Check 0 - 13 provides 14 brilliant colors on 1 paper giving instant pH checks over the entire range with accurate, reproducible readings; and immediate response. Available in different widths to meet your special requirements and preferences. Paper rolls come with dispenser; refills do not.
Product Family Options
Informations sur le produit
- DésignationLongueurLargeurpH RangeAvailabilityPrice
- pH paper jumbo dispenser Insta-Check15 ft.⁵⁄₃₂"0 - 13Each$51.90Carton of 10$458.65Supplier Number: 3015
- pH paper roll Insta-Check50 ft.½"0 - 13Each$33.90Case of 10$298.55Supplier Number: JR113
- pH paper roll Insta-Check50 ft.½"0 - 13Each$40.30Carton of 10$364.60Supplier Number: HJ613