Inside/Outside Vernier Caliper, Shiv Dial Sud & Sons
Catalogue # 470148-916
Fournisseur: Shiv Dial Sud & Sons
Spécifications
- Cat. no.:470148-916
- Measurement Range:0-5.5"/14 cm
- Accuracy:0.007"/0.1 mm
- Supplier No.:CM2027
Spécifications
À propos de cet article
Precise Vernier Caliper
- Measures inside and outside dimensions
- Uses metric and English units
- Thumb wheel for fine adjustments
- Locking mechanism