Insect Cornea Slide
Insect Cornea, Whole mount, Unstained
Catalog # 470177-532
Supplier: Avantor
Jump to:
Product Details & Documents
Illustrates Features of the Compound Eye
- Identifying features clearly distinguishable
- Rigorous quality control standards
Horsefly. Flat mount showing facets. Unstained.
Spécifications
- Désignation:Insect Cornea
- Stain:Unstained
- Orientation:Whole mount