Insect Compound Eye Slide
Insect Compound Eye, Section, Mason
  470181-638
 :   Avantor
Insect Compound Eye Slide
  470181-638
 :  Avantor
Insect Compound Eye Slide
Insect Compound Eye Slide
Insect Compound Eye, Section, Mason
$39.33
Each
Demonstrates Structures of the Compound Eye
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Radial section of compound eye showing cornea, rods, and cones.
  • Désignation:
    Insect Compound Eye
  • Stain:
    Mason
  • Orientation:
    Section