Innoculating Loops | Avantor
Innoculating Loops
Innoculating Loops
Certificates
Innoculating Loops
Innoculating Loops
3 Options Available Below
Product Details & Documents
Color set of four inoculating loops with brass handles and four colors.
  • Non-insulated aluminum handle
Twisted 3" lengths of non-calibrated 26 gauge nickel-chromium wire with 8 mm loops set in threaded aluminum handles. Wire is non-replaceable.
Product Family Options
Informations sur le produit
  • Désignation
    Matière
    Jauge
    Longueur
    Availability
    Price
  • Inoculating loop, 8 mm
    Aluminum
    26
    Handle: 8"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $9.65
    $9.65
    Each
       $9.65
    Supplier Number: INL703
  • Inoculating loop, 8 mm, set of 4
    Aluminum
    26
    Handle: 8"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $39.20
    $39.20
    Each
       $39.20
    Supplier Number: INL703-PK/4
  • Additional Wire loops
    -
    24
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 12
    Pack of 12 - $29.00
    $29.00
    Pack of 12
       $29.00
    Supplier Number: 3412