Ink
Ink, for Chromatography, Laboratory, 500 mL
  470006-536
 :  IX0061-500
Ink
  470006-536
 :  
Ink
Ink
Ink, for Chromatography, Laboratory, 500 mL
 :  
$36.00
Each
Shelf Life (months): 36
Storage: Green
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
 : This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
  • Désignation:
    Ink, for Chromatography
  • Taille:
    500 mL
  • Chemical/Physical Specifications:
    Black
  • Grade:
    Laboratory