ContactUsLinkComponent
Inertial Balance
  470149-084
undefined
Inertial Balance
  470149-084
 :  PH10590A
  • Description:
    Inertial Balance
  • Cat. no.:
    470149-084
  • Supplier No.:
    PH10590A
The oscillatory platform can carry various masses and is free to oscillate in a horizontal plane under the restoring forces of two straight-blade spring supports.
  • Economical
  • Solid Base Plate
  • Aluminum Trays
  • Instructions included
  • For Measuring Inertial Mass of Objects
It's used to measure the inertial mass of various objects.
The balance consists of separate aluminum trays securely connected by two spring blades.
 : This kit includes a base plate, a length of nylon cord, a cylindrical mass, and instructions.