Somso® Individual Human Bones
Supplier: MARCUS SOMMER
Jump to:
À propos de cet article
Replica bones are available for individual examination. Where noted natural bone is available.
- Somso replica bones
- Individual bones for comparison
- Five year manufacturers warranty
39 Options Available Below
Product Details & Documents
Replica bones are available for individual examination. Where noted natural bone is available.
- Somso replica bones
- Individual bones for comparison
- Five year manufacturers warranty
Product Family Options
Cranial
Informations sur le produit
- DésignationAvailabilityPrice
- Supplier Number: QS 69Spécifications:
More
Specifications
Cat. No.470030-004 - Discontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.470028-040 - Supplier Number: QS 9/3-16Spécifications:
More
Specifications
Cat. No.470030-044 - Supplier Number: QS 9/3-31Spécifications:
More
Specifications
Cat. No.470030-056 - Supplier Number: QS 9/3-25Spécifications:
More
Specifications
Cat. No.470030-052 - Supplier Number: QS 9/3-23Spécifications:
More
Specifications
Cat. No.470030-066 - Supplier Number: QS 9/3-11Spécifications:
More
Specifications
Cat. No.470030-046 - Supplier Number: QS 9/3-27Spécifications:
More
Specifications
Cat. No.470030-064 - Supplier Number: QS 9/3-14Spécifications:
More
Specifications
Cat. No.470030-048 - Supplier Number: QS 9/3-10Spécifications:
More
Specifications
Cat. No.470030-054 - Supplier Number: QS 9/3-12Spécifications:
More
Specifications
Cat. No.470030-050 - Supplier Number: QS 9/3-18Spécifications:
More
Specifications
Cat. No.470030-060 - Supplier Number: QS 9/3-29Spécifications:
More
Specifications
Cat. No.470030-062
Axial
Informations sur le produit
- DésignationAvailabilityPrice
- Supplier Number: QS 17/1Spécifications:
More
Specifications
Cat. No.470030-116 - Supplier Number: QS 17-CERVSpécifications:
More
Specifications
Cat. No.470030-098 - Supplier Number: QS19-LSpécifications:
More
Specifications
Cat. No.470217-492 - Supplier Number: QS19-RSpécifications:
More
Specifications
Cat. No.470217-494 - Supplier Number: QS 9/3-17Spécifications:
More
Specifications
Cat. No.470030-058 - Supplier Number: QS16/11-LSpécifications:
More
Specifications
Cat. No.470348-384 - Supplier Number: QS16/11-RSpécifications:
More
Specifications
Cat. No.470348-386 - Supplier Number: QS 17-LUMBSpécifications:
More
Specifications
Cat. No.470030-102 - Supplier Number: QS18-RSpécifications:
More
Specifications
Cat. No.470217-498 - Supplier Number: QS18-LSpécifications:
More
Specifications
Cat. No.470217-496 - Supplier Number: QS 17-THORSpécifications:
More
Specifications
Cat. No.470030-100
Appendicular
Informations sur le produit
- DésignationAvailabilityPrice
- Supplier Number: QS19/1-RSpécifications:
More
Specifications
Cat. No.470217-522 - Supplier Number: QS19/1-LSpécifications:
More
Specifications
Cat. No.470217-520 - Supplier Number: QS19/6-LSpécifications:
More
Specifications
Cat. No.470217-528 - Supplier Number: QS19/6-RSpécifications:
More
Specifications
Cat. No.470217-530 - Supplier Number: QS 19/10Spécifications:
More
Specifications
Cat. No.470218-978 - HumerusDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.470030-156 - Supplier Number: QS19/2-LSpécifications:
More
Specifications
Cat. No.470217-500 - Supplier Number: QS19/2-RSpécifications:
More
Specifications
Cat. No.470217-502 - Supplier Number: QS19/7-LSpécifications:
More
Specifications
Cat. No.470217-524 - Supplier Number: QS19/7-RSpécifications:
More
Specifications
Cat. No.470217-526 - Supplier Number: QS 19/3Spécifications:
More
Specifications
Cat. No.470030-150 - Supplier Number: QS19/9-LSpécifications:
More
Specifications
Cat. No.470217-504 - Supplier Number: QS19/9-RSpécifications:
More
Specifications
Cat. No.470217-506 - ScapulaDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.470030-142 - Supplier Number: QS19/5-LSpécifications:
More
Specifications
Cat. No.470217-532 - Supplier Number: QS19/5-RSpécifications:
More
Specifications
Cat. No.470217-534 - Supplier Number: QS19/8-LSpécifications:
More
Specifications
Cat. No.470217-508 - Supplier Number: QS19/8-RSpécifications:
More
Specifications
Cat. No.470217-510