Indicator Bottles
Bottle, Indicator, 60mL
Catalog # 470145-226
Supplier: Bel-Art
Supplier #: F11662-0000_SE
Jump to:
Product Details & Documents
Double density polyethylene bottles do double duty as a dropper bottle or as a spray bottle. Excellent for use with solutions or dyes.
Packaging: Package of 12.
Specifications
- Désignation:Bottle, Indicator, 60mL