Ward's® Igneous Rocks Poster
Catalogue # 470013-922
Fournisseur: Avantor
Spécifications
- Description:WARD’S Igneous Rocks Poster
- Cat. no.:470013-922
- Supplier No.:WARD470013-922
Spécifications
À propos de cet article
Depict the story of igneous rocks as never before with this colorful and dramatic poster.
- Size: 34W×21" H
Supplemental information includes a classification chart of common igneous rock types, graphic illustrations depicting common igneous rock structures, and Bowen’s reaction series.
Poster can also be used as a supplement to the WARD’S Rock Cycle CD-ROMs.