Ward's® Igneous Rock Collection
Ward's® Igneous Rock Collection
Supplier:   Avantor
Ward's® Igneous Rock Collection
Supplier:  Avantor
Certificates
Ward's® Igneous Rock Collection
Ward's® Igneous Rock Collection
2 Options Available Below
Product Details & Documents
Samples of Intrusive and Extrusive Igneous Rocks
  • Introduce concepts of the Earth's crust
  • Compartmented box
Provide your students with an excellent introduction to the forces at work beneath the Earth’s crust. The set is housed in a compartmented collection box and includes an identification list.
Contents:
12 numbered samples, 1" x 2"
Rocks:
basalt, diorite, gabbro, granite, obsidian, pegmatite, peridotite, pumice, rhyolite (2), scoria, syenite
Hand Specimen Set:
Large hand samples are ideal for demonstration and display. This set is housed in individual, labeled plastic display trays and comes with an identification list.
Contents:
12 numbered samples, 3" x 4"
Product Family Options
Product Information
  • Désignation
    No. of Samples
    Availability
    Price
  • Igneous Rock Collection
    12
    Log in for availability
    Log in for availability
    Each
    Each - $48.99
    $48.99
    Each
       $48.99
  • Igneous Rock Collection, Hand Specimen Set
    12
    Log in for availability
    Log in for availability
    Each
    Each - $277.25
    $277.25
    Each
       $277.25