Ice Cream Making Kit: A Heat Transfer Experiment
470104-476
: Avantor
- Description:Ice Cream Making Kit
- No. of Lab Groups:5
- Cat. no.:470104-476
- Supplier No.:WARD470104-476
Science Has Never Been Sweeter!
- Perfect for Grades 4-9
- Instruction and Lesson Plans Included
Using this kit, students use a simple technique to create edible ice cream and learn about heat transfer, phase change, and insulation along the way. The entire activity, from heat transfer to clean-up, can be performed in a single 45-minute lab period. Kit includes enough materials for five lab stations of six students per group, with 20 ice cream recipes included. Each recipe yields approximately 2-3 cups of ice cream. We recommend using a glue gun to apply the pre-cut insulation to the reusable cans.
Includes:
Water Softener Salt
Metal Cans with Lid (2 sizes)
Foam Insulation
Duct Tape
Instructions
Required but Not Provided: Ice, Glue Gun, Measuring Cup or Beakers, Bowls or Cups, Spoons, Blender (optional).