Eisco® Digestive System
Digestive System, 2, 30 cm, 81 cm, 7 cm
Catalog # 470111-070
Supplier: Wards
Fournisseur #: AM0093
Jump to:
Product Details & Documents
Explore the full digestive system.
- Mounted on board
- Hand painted
- Includes key card
Model of human digestive system showing nose, mouth cavity and pharynx, esophagus, gastrointestinal tract, liver with gall bladder, pancreas and spleen, duodenum, cecum, and rectum are opened. Transverse colon is removable to show internal structure. Excellent tool for showing path of food.
Spécifications
- Désignation:Digestive System
- Hauteur:7 cm
- Longueur:81 cm
- Width:30 cm
- No. of Parts:2
Accessories
Informations sur le produit
- Catalog #No. of PartsWidthAvailabilityPrice
- Digestive system