ContactUsLinkComponent
How Good Is Your Suntan Lotion? Kit
How Good Is Your Suntan Lotion? Kit
Catalog # 470200-494
Supplier:  Kemtec
undefined
How Good Is Your Suntan Lotion? Kit
Catalog # 470200-494
Supplier:  Kemtec
Supplier Number:  4-218
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    How Good Is Your Suntan Lotion? Kit
  • Cat. no.:
    470200-494
  • Supplier No.:
    4-218
Specifications
About this item
Do commercial sunscreens really work? Test them and find out with this kit.
It makes an excellent class study but is also a good basis for individual study. The kit is designed for 24 students working in groups of four.
Delivery: Includes materials for six student groups, student manuals, student data sheets, assessment materials, special needs materials, and an instructor’s manual.