Hooked Mass Set
Mass Set, Hooked, Economy, Set/20, 1g - 1kg
Catalog # 470007-384
Supplier: Sempco
Fournisseur #: WS-9109
Jump to:
Product Details & Documents
This hooked mass set is an ideal addition to your classroom - its economical, convenient, and durable! A set of twenty plated steel hex weights with hook, ranging from 1g to 1kg, conveniently stored on a fitted wooden base economically priced.
Simply bring the storage base to the experiment site and all the weights will be at your disposal! The Hooked Mass Set offers a great way to demonstrate weight at any grade level.
Exact masses included in this set are: One of each of 1g, 2g, 3g, 4g, 5g, 6g, 7g, 8g, 9g, 20g, 30g, 40g, 50g, 250g, 500g, and 1kg. As well as two each of 10g and 100g.
Spécifications
- Désignation:Mass Set, Hooked, Economy, Set/20, 1g - 1kg