Hooks Collars
Hooks Collars
Supplier:   GSC International
Hooks Collars
Supplier:  GSC International
Certificates
Hooks Collars
4 Options Available Below
Product Details & Documents
The Hooks Collar is constructed with a ring that fits over a support stand rod, a hook and thumb screw.
  • Metal ring
  • Metal hook
  • Different sizes and materials to fit all classroom needs and budgets
The Hooks Collar is used in a physics classroom for suspending items such as pulleys, cords, or weights from a rod or crass bar.
Caution: Prop 65 Lead Warnings are attached to the package.
Product Family Options
Product Information
  • Désignation
    Matière
    Taille
    Availability
    Price
  • Hooks collar
    Metal
    13 mm Int.Ø
    Log in for availability
    Log in for availability
    Each
    Each - $10.75
    $10.75
    Each
       $10.75
    Supplier Number: HKCLR-13
  • Hooks collar
    Plastic
    13 mm Int.Ø
    Log in for availability
    Log in for availability
    Each
    Each - $5.70
    $5.70
    Each
       $5.70
    Supplier Number: HKCLR-13-C
  • Hooks collar
    Metal
    19 mm Int.Ø
    Log in for availability
    Log in for availability
    Each
    Each - $5.70
    $5.70
    Each
       $5.70
    Supplier Number: HKCLR-19
  • Hooks collar
    Plastic
    19 mm Int.Ø
    Log in for availability
    Log in for availability
    Each
    Each - $5.85
    $5.85
    Each
       $5.85
    Supplier Number: HKCLR-19-C