Hook And Chain Set
Hook And Chain, Stainless steel, 152 mm (6")
Catalog # 470007-570
Supplier: Dunrite Instruments
Fournisseur #: 141130
Jump to:
Product Details & Documents
Three stainless steel hooks chained to a loop hold large specimens in place when dissecting.
Spécifications
- Désignation:Hook And Chain
- Longueur:152 mm (6")
- Matière:Stainless steel