High Form Porcelain Crucibles
Supplier: SILICO & CHEMICO PORCELAIN SE
Certificates
About this item
These crucibles are similar to wide-form porcelain crucibles but taller.
- Without covers
- Max. use temp. is 1050 Celsius
- Rate of cooling and heating should not increase 200 degrees per hour
They don't have covers.
Frequently Bought Together