Weighing Bottles | Avantor
Weighing Bottles
Flint glass weighing bottle with ground glass stopper, 20 ml, 30 mm, 50 mm
Catalog # 470235-764
Supplier:   GSC International
Fournisseur #:  WB4132-20
Weighing Bottles
Catalog # 470235-764
Supplier:  GSC International
Weighing Bottles
Weighing Bottles
Flint glass weighing bottle with ground glass stopper, 20 ml, 30 mm, 50 mm
Price
5,20 $
Log in pour la disponibilité
Each Pack of 20
Product Details & Documents
Designed for use in the lab to take precise weight of solids.
  • Flint glass
  • Ground glass joints
  • Consistent weight
Flint glass weighing bottles feature a ground glass stopper. Available in two sizes.
Spécifications
  • Ø:
    30 mm
  • Capacité:
    20 ml
  • Désignation:
    Flint glass weighing bottle with ground glass stopper
  • Hauteur:
    50 mm