ContactUsLinkComponent
Herbarium Mounting Paper
Catalogue # 470106-370
Fournisseur:  xpedx
undefined
Herbarium Mounting Paper
Catalogue # 470106-370
Fournisseur:  xpedx
Supplier Number:  10002904
Spécifications
  • Description:
    Herbarium Mounting Paper
  • Thickness:
    0.0105"
  • Cat. no.:
    470106-370
  • Supplier No.:
    2261169
Spécifications
À propos de cet article
Make your herbarium mounts last with this top-quality stock. The white paper has a high rag content that is ideal for mounting purposes. Size: 29 x 42cm. Package of 100. Thickness of .0105 inches.