Heavy-Duty Metal Service Cart
470020-364
: Lakeside
- Description:Heavy-Duty Service Cart
- Cat. no.:470020-364
- Supplier No.:422
The stainless steel construction is durable enough for heavy laboratory loads. It also features 4" swivel casters.
Carrying capacity: 500 lbs.
Size: 30 1/2"L x 17 3/4"W x 32 1/4"H