Heat Transfer Activity | Avantor
Heat Transfer Activity
Supplier:   Avantor
Heat Transfer Activity
Supplier:  Avantor
Certificates
Heat Transfer Activity
Heat Transfer Activity
1 Options Available Below
Product Details & Documents
Apparatus Shows Effect of Heated Water on Other Materials
  • Literature included
  • Observe transfer of heat under different conditions
Heat energy cannot be seen, but with this calorimeter featuring an aluminum transfer bar, students can observe heat transfer under a variety of different conditions by measuring the temperatures of the insulated containers before and after connecting them with the metal bar. You can also vary the experiment by using water of different temperatures, or unequal amounts of liquid.
Ordering information: Please note, lids are not included with replacement containers. Materials included are Calorimeters, Aluminum transfer bar, thermometers. Materials Required but not Included: Boiling water, Wood Alcohol, 100 mL Graduated cylinder, Timer/stopwatch.
Product Family Options
Informations sur le produit
  • Désignation
    No. of Lab Groups
    Availability
    Price
  • Heat Transfer Lab Activity
    15
    Discontinued
  • Heat Transfer Activity
    1
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $35.65
    $35.65
    Liquidation
    Each
       $35.65
  • Replacement Insulated Containers
    Discontinued