ContactUsLinkComponent
Hand Pump with Vacuum Plate
Catalogue # 470121-430
Fournisseur:  Wards
undefined
Hand Pump with Vacuum Plate
Catalogue # 470121-430
Fournisseur:  Wards
Supplier Number:  PH0166B
Spécifications
  • Description:
    Hand Pump with Vacuum Plate
  • Cat. no.:
    470121-430
  • Supplier No.:
    PH0166B
Spécifications
À propos de cet article
Efficient combination of a pump and a vacuum plate.
  • Plate features a silicon seal, which allows perfect sealing of the bell jar with the plate
  • 20 cm diameter size
Just a few strokes of the pump are enough to establish a vacuum in a bell jar, so students can observe the reducing sound level of the bell. The plate can also be used with electric vacuum pumps with the help of a three-way stopcock under the plate.