To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:WARD’S Gunshot Residue Presumptive Test Kit
- No. of Lab Groups:15
- Cat. no.:470024-240
- Supplier No.:DD6002-KIT
Specifications
About this item
Colorimetric test for trace evidence.
- Grades 9-12
- Estimated class/lab time required: 30 minutes
This simple, two-part presumptive test will help you quickly determine whether or not a surface has recently been exposed to a discharged firearm. Simply wipe down the surface in question, add the chemical reagents, and observe the results. A rapid color-change takes place to verify the presence of nitrates, the main component of gunpowder residue (GSR), while a different color change will verify the presence of heavy metals associated with GSR. Each kit contains complete instructions and enough materials to perform 30 tests.