Green Plants Microslide | Avantor
Green Plants Microslide
Green Plants Microslide
Certificates
Green Plants Microslide
Green Plants Microslide
2 Options Available Below
Product Details & Documents
Examine Different Plants and their Characteristics
  • Arrows help students locate important features
  • Mounted in a clear plastic holder that protects it on both sides
Microslides consist of a strip of film with photomicrographs, each reproduced exactly as you would see them through a professional microscope under ideal conditions. You can supply all your students with their own Microslides. A detailed text folder is included. Viewers are available separately.
Ordering information: Set includes Green leaf (cs, 110X), Green leaf (cs, 350X), Stomate (cs, 480X), Root hairs (wm, 35X), Root (cs, 20X), Celery (Is, 40X), Corn (cs, 145X), Veins in a leaf (wm, 40X), Chloroplasts; The food factory (30,000X), Leaf with starch (wm, 35X), and Bean seed (wm, 2X).
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Microslide, green plants set
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $19.60
    $19.60
    Pack of 1
       $19.60
    Supplier Number: T-119
  • Microslide, green plants set, set of 15
    Log in pour la disponibilité
    Log in pour la disponibilité
    Box of 15
    Box of 15 - $292.00
    $292.00
    Box of 15
       $292.00
    Supplier Number: T-119-15-B