Ward's® Granite (Gray) | Avantor
Ward's® Granite (Gray)
Supplier:   Avantor
Ward's® Granite (Gray)
Supplier:  Avantor
Certificates
Ward's® Granite (Gray)
Ward's® Granite (Gray)
4 Options Available Below
Product Details & Documents
Fine to medium-grained with biotite.
  • Collected in Vermont
  • Multiple sizes and pack options to fit your specific needs
  • Cut by Ward's geologists to guarantee top market quality
  • Matching thin section available
Student Specimens - Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.
Hand Specimens - Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.
Bulk Pack - Includes 1 kilo of assorted pieces (quantity varies). Assorted pieces are 1.25×1.25" to 2×3". Packaged in a labeled storage box for easy organization.
All specimen dimensions are considered an average size range.
Product Family Options
Specimens
Informations sur le produit
  • Désignation
    Specimen Dimensions
    Availability
    Price
  • Granite (Gray), Student Specimens
    1×1" - 1×1"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $27.95
    $27.95
    Pack of 10
       $27.95
  • Granite (Gray), Hand Specimen
    2×3" - 3×4"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $28.60
    $28.60
    Each
       $28.60
  • Granite (Gray), Bulk Pack, 1 kg
    Size will vary
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $29.70
    $29.70
    Each
       $29.70
  • Granite (Gray) thin section
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $96.59
    $96.59
    Each
       $96.59
Storage
Informations sur le produit
  • Catalog #
    Désignation
    Availability
    Price
  • Storage box
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2.35
    $2.35
    Each
       $2.35