Gonium Slide
Gonium, Whole mount, Fuchsin and Fast Green
Catalog # 470180-976
Supplier: Avantor
Jump to:
Product Details & Documents
Investigate Algae Structure
- Identifying features clearly distinguishable
- Rigorous quality control standards
Colonial motile algae, showing cytological detail.
Spécifications
- Désignation:Gonium
- Stain:Fuchsin and Fast Green
- Orientation:Whole mount