Gonionemus | Avantor
Gonionemus
Gonionemus, Preserved, Jar of 10
Catalog # 470001-204
Supplier:   Avantor
Gonionemus
Catalog # 470001-204
Supplier:  Avantor
Gonionemus
Gonionemus
Gonionemus, Preserved, Jar of 10
Price
$55.93
Log in pour la disponibilité
Pack of 10
Product Details & Documents
Formalin Preserved
This small cosmopolitan jellyfish clearly shows the structure of hydrozoan medusae. Size: 3/8".
Spécifications
  • Désignation:
    Gonionemus, Preserved, Jar of 10
Accessories
Informations sur le produit
  • Catalog #
    Grade
    Size
    Availability
    Price
  • WARDSafe™
    WARDSafe™
    470233-546
    Laboratory
    19 L
    Log in pour la disponibilité
    Each (19l) - $345.85