Gold Pans
Catalog # 470144-648
Supplier: Estwing Manufacturing
Specifications
- Description:Gold Pans, 12" Dia., 1 lb.
- Cat. no.:470144-648
- Supplier No.:12-12IN
Specifications
About this item
Rugged Construction Withstands Years of Use
Heavy-gauge steel pans with ribbed rims stand up to the elements as well as geologists and prospectors separating gold or other heavy minerals from stream gravel. The pans come in two sizes for processing smaller or larger quantities.