Glycogen Slide
Glycogen, Section, Periodic acid Schiff reaction
Catalog # 470182-594
Supplier: Avantor
Jump to:
Product Details & Documents
Illustrates Energy Storage
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal liver. Glycogen granules stained red and nucleus blue. Compare to Glycogen Removed Slide.
Spécifications
- Désignation:Glycogen
- Stain:Periodic acid Schiff reaction
- Orientation:Section