Gloeocapsa Slide | Avantor
Gloeocapsa Slide
Gloeocapsa Slide, Whole Mount, Safranin
Catalog # 470177-416
Supplier:   Avantor
<i>Gloeocapsa</i> Slide
Catalog # 470177-416
Supplier:  Avantor
<i>Gloeocapsa</i> Slide
Gloeocapsa Slide
Gloeocapsa Slide, Whole Mount, Safranin
Price
$8.95
Log in pour la disponibilité
Each
Product Details & Documents
Cyanobacteria sheaths.
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Individuals and colonies in gelatinous capsules.
Spécifications
  • Désignation:
    Gloeocapsa Slide
  • Stain:
    Safranin
  • Orientation:
    Whole Mount