GLOBE Cloud Chart
GLOBE Cloud Chart
Catalog # 470104-448
Supplier: Peco Sales
Supplier #: 33485
Jump to:
Product Details & Documents
Clouds cover the world and this chart, written in English, French, Spanish, Arabic, Chinese and Russian make that point very well indeed.
- Size: 11×15"
Provides descriptions and visual examples of the ten basic cloud types: cirrus, cirrostratus, cirrocumulus, altostratus, altocumulus, cumulus, nimbostratus, stratus, cumulonimbus, and stratocumulus.
Specifications
- Désignation:GLOBE Cloud Chart