Gilley Induction Coil Set
Gilley Induction Coil Set
470005-394
: Wards
: GLCL
An excellent set of materials for demonstrating electromagnetism and induced currents, the set includes two electromagnetic coils mounted on plastic saddle blocks, round and square iron cores, split round iron cores, a double core, and a card for showing lines of force. Set of two.
- Désignation:Gilley Induction Coil Set