General Botany Slide Sets
Catalogue # 470177-794
Fournisseur: Avantor
Spécifications
- Type:Plant
- Description:General Botany Slide Sets
- No. of Slides:25
- Cat. no.:470177-794
- Supplier No.:WARD470177-794
Spécifications
À propos de cet article
Using these sets, students compare and contrast botanical life forms.
- Rigorous Quality Control Standards
- Clearly Distinguishable Identifying Features
- Twenty-Five Examples of Botanical Life Forms
These sets have an outstanding selection with 25 examples of bacteria, algae, fungi, bryophytes, ferns, gymnosperms, stems, leaves, flower structures, and monocot/dicot roots.
The identifying features of these examples are clearly visible on the slides.