Gas Turbine Engine Model
Catalog # 470121-112
Supplier: Wards
Specifications
- Description:Gas Turbine Engine Model
- Cat. no.:470121-112
- Supplier No.:PH0494
Specifications
About this item
Hands-On Visual Aid
- Model is made of light metal and provided on base
This section cut model illustrates multiple engine concepts and characteristics. Demonstrate air intake, axial flow, double stage compressor, fuel supply, combustion chamber, turbine rotor, jet thrust, exhaust, and more.