Gammarus Slide
Gammarus, Whole mount, Schneider's and Fast Green
Catalog # 470181-594
Supplier: Avantor
Jump to:
Product Details & Documents
Common Amphipod Crustacean Genus
- Identifying features clearly distinguishable
- Rigorous quality control standards
An example of the structure of a typical amphipod.
Spécifications
- Désignation:Gammarus
- Stain:Schneider's and Fast Green
- Orientation:Whole mount