Gallbladder, Mammal Slide
Gallbladder, Mammal, Section, Hematoxylin and Eosin
Catalog # 470182-888
Supplier: Avantor
Jump to:
Product Details & Documents
Digestive Organ
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal. Through mucosa with simple columnar epithelium, serous layer, and muscular layer.
Specifications
- Désignation:Gallbladder, Mammal
- Stain:Hematoxylin and Eosin
- Orientation:Section