Ward's® Gabbro
Supplier:   Avantor
Ward's® Gabbro
Supplier:  Avantor
Certificates
Ward's® Gabbro
4 Options Available Below
Product Details & Documents
Coarse-grained plagioclase and pyroxene with some olivine.
  • Collected in Minnesota
  • Multiple sizes and pack options to fit specific needs
  • Cut by Ward's geologists to guarantee top market quality
  • Matching thin section available
Student Specimens – Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.
Hand Specimens – Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.
Bulk Pack – Includes 1 kilo of assorted pieces (quantity varies). Assorted pieces are 1.25×1.25" - 2×3". Packaged in a labeled storage box for easy organization.
All Specimen dimensions are considered an average size range.
Product Family Options
Specimens
Product Information
  • Désignation
    Specimen Dimensions
    Availability
    Price
  • Gabbro thin section
    Log in for availability
    Log in for availability
    Each
    Each - $96.59
    $96.59
    Each
       $96.59
  • Gabbro, bulk pack, 1 kg
    Size will vary
    Log in for availability
    Log in for availability
    Each
    Each - $36.95
    $36.95
    Each
       $36.95
  • Gabbro, hand specimen
    2×3" - 3×4"
    Log in for availability
    Log in for availability
    Each
    Each - $36.95
    $36.95
    Each
       $36.95
  • Gabbro, student specimens
    1×1 - 1×1"
    Log in for availability
    Log in for availability
    Pack of 10
    Pack of 10 - $36.00
    $36.00
    Pack of 10
       $36.00
Storage
Product Information
  • Catalog #
    Désignation
    Availability
    Price
  • Storage box
    Log in for availability
    Log in for availability
    Each
    Each - $2.35
    $2.35
    Each
       $2.35