Funnel Support with Base
Funnel Support with Base, 11 x 38 cm (Base), 43 cm
Catalog # 470005-858
Supplier: Wards
Fournisseur #: CH0689C
Jump to:
Product Details & Documents
Made from fine, oil finished hardwood designed to support up to four funnels. The height is adjustable and the holes are slotted for easy placement of funnels. The base measures 11 x 38 cm.
Spécifications
- Désignation:Funnel Support with Base
- Dimensions:11 x 38 cm (Base)
- Hauteur:43 cm