Funnel Sets | Avantor
Funnel Sets
Supplier:   Avantor
Funnel Sets
Supplier:  Avantor
Certificates
Funnel Sets
2 Options Available Below
Product Details & Documents
Each set includes three funnels of different sizes. Small funnels are made of polypropylene; large funnels are made of polyethylene. Sizes in small funnel set: 60 mm top dia. x 20 mm stem, 80 mm top dia. x 22 mm stem, 110 mm top dia. x 36 mm stem. Sizes in large set: 127 mm top dia. x 45 mm stem, 160 mm top dia. x 50 mm stem, 200 mm top dia. x 60 mm stem.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Funnel Set, Set of Three
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 3
    Pack of 3 - $19.10
    $19.10
    Log in pour la disponibilité
    Case of 216
    Case of 216 - $1,327.35
    $1,327.35
    Pack of 3
       $19.10
    Case of 216
       $1,327.35
    Supplier Number: 248
  • Funnel set, large, HDPE, set of three
    Log in pour la disponibilité
    Log in pour la disponibilité
    Kit of 3
    Kit of 3 - $64.80
    $64.80
    Kit of 3
       $64.80
    Supplier Number: -