Funnel Set, Dynalon | Avantor
Funnel Set, Dynalon
Funnel Set
Catalog # 470146-840
Supplier:   Dynalon
Fournisseur #:  241645-0SET
Funnel Set, Dynalon
Catalog # 470146-840
Supplier:  Dynalon
Funnel Set, Dynalon
Funnel Set, Dynalon
Funnel Set
Price
$133.15
Log in pour la disponibilité
Kit of 6
Product Details & Documents
Two each of top diameter 65mm and 100mm and 120mm. All are 60° angle. Set of six.
Spécifications
  • Désignation:
    Funnel Set