Full Spear Point Dissecting Needle
Full Spear Point Dissecting Needle, Stainless steel, 152 mm (6"), Spear
Catalog # 470018-970
Supplier: Dunrite Instruments
Fournisseur #: 141035
Jump to:
Product Details & Documents
The spear point is useful for cutting fascia and cleaning tissue from small structures.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Spécifications
- Désignation:Full Spear Point Dissecting Needle
- Longueur:152 mm (6")
- Matière:Stainless steel
- Tip Style:Spear